siRNA Details
Gene Symbol: Jag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240666.2 | XP_006240728.1 | 11 | 1544 | ||
XM_006240667.2 | XP_006240729.1 | 10 | 1429 | |||
GenBank mRNA | U70050.1 | - | 11 | 1356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
