siRNA Details
Gene Symbol: Vtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019156.2 | NP_062029.2 | 4 | 624 | ||
GenBank mRNA | AY318963.1 | - | 612 | |||
BC105821.1 | - | 4 | 624 | 7371104 | ||
FQ209609.1 | - | 4 | 636 | |||
FQ209682.1 | - | 4 | 629 | |||
FQ210146.1 | - | 2 | 244 | |||
FQ210518.1 | - | 4 | 629 | |||
FQ212575.1 | - | 4 | 623 | |||
FQ218318.1 | - | 4 | 636 | |||
FQ218401.1 | - | 2 | 197 | |||
FQ219297.1 | - | 4 | 637 | |||
U44845.1 | - | 4 | 614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
