siRNA Details
Gene Symbol: Aqp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022181.1 | NP_071517.1 | 4 | 907 | ||
GenBank mRNA | AF083879.1 | - | 4 | 907 | ||
L28113.1 | - | 4 | 935 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
