siRNA Details
Gene Symbol: Penk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017139.1 | NP_058835.1 | 2 | 1082 | ||
XM_006237835.1 | XP_006237897.1 | 3 | 935 | |||
GenBank mRNA | BC083563.1 | - | 2 | 1073 | 7112833 | |
FQ213928.1 | - | 3 | 915 | |||
M28263.1 | - | 3 | 803 | |||
S49491.1 | - | 3 | 873 | |||
Y07503.1 | - | 2 | 1082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
