siRNA Details
Gene Symbol: Tyms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019179.1 | NP_062052.1 | 4 | 465 | ||
XM_008767420.1 | XP_008765642.1 | 4 | 954 | |||
XM_008767421.1 | XP_008765643.1 | 5 | 510 | |||
GenBank mRNA | BC126093.1 | - | 4 | 479 | 8362547 | |
L12138.1 | - | 4 | 465 |

More Information
Other Species Targeted

Gene Ontology Categories:
