siRNA Details
Gene Symbol: Tpsb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019180.2 | NP_062053.2 | 3 | 315 | ||
GenBank mRNA | D38455.1 | - | 4 | 325 | ||
U67909.1 | - | 3 | 316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
