siRNA Details
Gene Symbol: Rpl23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007599.2 | NP_001007600.1 | 3 | 235 | ||
GenBank mRNA | BC058500.1 | - | 3 | 239 | 6921061 | |
FQ216928.1 | - | 3 | 241 | |||
FQ221252.1 | - | 3 | 241 | |||
FQ221690.1 | - | 3 | 265 | |||
FQ222524.1 | - | 3 | 241 | |||
FQ222678.1 | - | 3 | 241 | |||
FQ223359.1 | - | 3 | 241 | |||
FQ224963.1 | - | 3 | 241 | |||
FQ228675.1 | - | 3 | 242 | |||
X58200.1 | - | 3 | 233 |

More Information
Other Species Targeted

Gene Ontology Categories:
