siRNA Details
Gene Symbol: Rps3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017153.1 | NP_058849.1 | 2, 3 | 193 | ||
GenBank mRNA | BC058483.1 | - | 2, 3 | 198 | 6887884 | |
FQ209727.1 | - | 2, 3 | 193 | |||
FQ211699.1 | - | 2, 3 | 194 | |||
FQ211708.1 | - | 2, 3 | 193 | |||
FQ216850.1 | - | 2, 3 | 193 | |||
FQ217809.1 | - | 2, 3 | 193 | |||
FQ219020.1 | - | 2, 3 | 193 | |||
FQ219404.1 | - | 2, 3 | 192 | |||
FQ219522.1 | - | 2, 3 | 194 | |||
FQ219543.1 | - | 2, 3 | 194 | |||
FQ219867.1 | - | 2, 3 | 194 | |||
FQ220002.1 | - | 2, 3 | 193 | |||
FQ220027.1 | - | 2, 3 | 193 | |||
FQ220037.1 | - | 2, 3 | 194 | |||
FQ220045.1 | - | 2, 3 | 193 | |||
FQ220135.1 | - | 2, 3 | 194 | |||
FQ220260.1 | - | 2, 3 | 193 | |||
FQ220261.1 | - | 2, 3 | 194 | |||
FQ220317.1 | - | 2, 3 | 193 | |||
FQ220504.1 | - | 2, 3 | 193 | |||
FQ220529.1 | - | 2, 3 | 193 | |||
FQ220577.1 | - | 2, 3 | 194 | |||
FQ220731.1 | - | 2, 3 | 193 | |||
FQ221185.1 | - | 2, 3 | 194 | |||
FQ221431.1 | - | 2, 3 | 194 | |||
FQ221460.1 | - | 2, 3 | 193 | |||
FQ221744.1 | - | 2, 3 | 193 | |||
FQ221931.1 | - | 2, 3 | 193 | |||
FQ222056.1 | - | 2, 3 | 193 | |||
FQ222736.1 | - | 2, 3 | 194 | |||
FQ222773.1 | - | 2, 3 | 194 | |||
FQ222846.1 | - | 2, 3 | 223 | |||
FQ222869.1 | - | 2, 3 | 193 | |||
FQ222876.1 | - | 2, 3 | 193 | |||
FQ223535.1 | - | 2, 3 | 200 | |||
FQ223540.1 | - | 2, 3 | 193 | |||
FQ225080.1 | - | 2, 3 | 194 | |||
FQ225145.1 | - | 2, 3 | 193 | |||
FQ225713.1 | - | 2, 3 | 192 | |||
FQ227696.1 | - | 2, 3 | 193 | |||
FQ227805.1 | - | 2, 3 | 194 | |||
FQ227878.1 | - | 2, 3 | 194 | |||
FQ228312.1 | - | 2, 3 | 194 | |||
FQ228317.1 | - | 2, 3 | 193 | |||
FQ228376.1 | - | 2, 3 | 194 | |||
FQ228558.1 | - | 2, 3 | 193 | |||
FQ229234.1 | - | 2, 3 | 193 | |||
FQ229381.1 | - | 2, 3 | 194 | |||
FQ229520.1 | - | 2, 3 | 193 | |||
FQ229870.1 | - | 2, 3 | 193 | |||
FQ229960.1 | - | 2, 3 | 193 | |||
FQ231162.1 | - | 2, 3 | 194 | |||
FQ231673.1 | - | 2, 3 | 193 | |||
FQ232329.1 | - | 2, 3 | 195 | |||
FQ232836.1 | - | 2, 3 | 193 | |||
FQ232928.1 | - | 2, 3 | 194 | |||
FQ233768.1 | - | 2, 3 | 197 | |||
FQ234311.1 | - | 2, 3 | 193 | |||
FQ234537.1 | - | 2, 3 | 193 | |||
M84716.1 | - | 2, 3 | 187 | |||
X75161.1 | - | 2, 3 | 193 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
