siRNA Details
Gene Symbol: Cyp2b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017156.1 | NP_058852.1 | 725 | |||
GenBank mRNA | X63545.1 | - | 725 |
Gene Symbol: Cyp2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134844.1 | NP_001128316.1 | 700 | |||
XM_008765491.1 | XP_008763713.1 | 719 | ||||
XM_008765492.1 | XP_008763714.1 | 719 | ||||
XM_008765493.1 | XP_008763715.1 | 1081 | ||||
GenBank mRNA | BC166502.1 | - | 700 | 7122207 | ||
J00719.1 | - | 668 | ||||
M37134.1 | - | 668 |
Gene Symbol: Cyp2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198676.1 | NP_001185605.1 | 5 | 683 | ||
GenBank mRNA | K01721.1 | - | 2 | 182 | ||
M34452.1 | - | 4 | 372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
