siRNA Details
Gene Symbol: Cyp2c7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017158.2 | NP_058854.1 | 9 | 1416 | ||
GenBank mRNA | BC097939.1 | - | 9 | 1399 | 7368637 | |
BC127503.1 | - | 9 | 1455 | 7371672 | ||
M18335.1 | - | 9 | 1417 | |||
M31031.1 | - | 9 | 1374 |
Gene Symbol: LOC100361547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759635.1 | XP_008757857.1 | 9 | 1272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
