siRNA Details
Gene Symbol: Prkce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017171.1 | NP_058867.1 | 13, 14 | 2258 | ||
GenBank mRNA | AY642593.1 | - | 13, 14 | 2072 | ||
M18331.1 | - | 13, 14 | 2258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
