siRNA Details
Gene Symbol: Fgf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133286.1 | NP_579820.1 | 3 | 199 | ||
XM_006231436.2 | XP_006231498.1 | 3 | 391 | |||
XM_006231437.2 | XP_006231499.1 | 4 | 319 | |||
XM_008760398.1 | XP_008758620.1 | 4 | 286 | |||
GenBank mRNA | AB079113.1 | - | 3 | 199 |

More Information
Other Species Targeted

Gene Ontology Categories:
