siRNA Details
Gene Symbol: Akt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031575.1 | NP_113763.1 | 4, 5 | 485 | ||
GenBank mRNA | D49836.1 | - | 4, 5 | 485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
