siRNA Details
Gene Symbol: Akt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031575.1 | NP_113763.1 | 4 | 464 | ||
GenBank mRNA | D49836.1 | - | 4 | 464 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011785, BC066861, AF124142.
Gene Ontology Categories:
