siRNA Details
Gene Symbol: Htt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024357.3 | NP_077333.2 | 33 | 4428 | ||
XM_006251230.2 | XP_006251292.1 | 32 | 4366 | |||
XM_006251232.2 | XP_006251294.1 | 33 | 4414 | |||
XM_008770274.1 | XP_008768496.1 | 31 | 3880 | |||
GenBank mRNA | U18650.1 | - | 33 | 4273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
