siRNA Details
Gene Symbol: Htt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024357.3 | NP_077333.2 | 48 | 6570 | ||
XM_006251230.2 | XP_006251292.1 | 47 | 6508 | |||
XM_006251232.2 | XP_006251294.1 | 48 | 6556 | |||
XM_008770274.1 | XP_008768496.1 | 46 | 6022 |

More Information
Other Species Targeted

Gene Ontology Categories:
