siRNA Details
Gene Symbol: LOC100910732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751066.2 | XP_003751114.1 | 12 | 1414 | ||
XM_006248473.1 | XP_006248535.1 | 13 | 1543 | |||
XM_008768778.1 | XP_008767000.1 | 12 | 2975 |
Gene Symbol: Pak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053306.3 | NP_445758.2 | 1266 | |||
GenBank mRNA | S80221.1 | - | 1185 | |||
U19967.1 | - | 1266 | ||||
U35345.1 | - | 1185 |

More Information
Other Species Targeted

Gene Ontology Categories:
