siRNA Details
Gene Symbol: Msra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053307.2 | NP_445759.1 | 6 | 638 | ||
GenBank mRNA | AY005464.1 | - | 6 | 628 | ||
BC087009.1 | - | 6 | 638 | 7107102 | ||
DQ989019.1 | - | 7 | 670 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
