siRNA Details
Gene Symbol: Atp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013795.5 | NP_038823.2 | 2 | 219 | ||
GenBank mRNA | AK005175.1 | - | 2 | 166 | ||
AK005321.1 | - | 2 | 196 | |||
AK005380.1 | - | 2 | 197 | |||
AK008002.1 | - | 2 | 156 | |||
AK008012.1 | - | 2 | 193 | |||
AK008030.1 | - | 2 | 182 | |||
AK008036.1 | - | 2 | 196 | |||
AK008087.1 | - | 2 | 170 | |||
AK008169.1 | - | 2 | 196 | |||
AK008183.1 | - | 2 | 184 | |||
AK008251.1 | - | 2 | 184 | |||
AK008321.1 | - | 2 | 192 | |||
AK009465.1 | - | 2 | 185 | |||
AK009468.1 | - | 2 | 185 | |||
AK009471.1 | - | 2 | 185 | |||
AK009612.1 | - | 2 | 178 | |||
AK009806.1 | - | 2 | 184 | |||
AK011038.1 | - | 2 | 163 | |||
AK011046.1 | - | 2 | 163 | |||
AK011099.1 | - | 2 | 190 | |||
AK011842.1 | - | 2 | 156 | |||
AK019252.1 | - | 2 | 185 | |||
AK019274.1 | - | 2 | 185 | |||
AK019320.1 | - | 2 | 192 | |||
AK019343.1 | - | 2 | 156 | |||
AK019350.1 | - | 2 | 184 | |||
AK019355.1 | - | 2 | 184 | |||
AK146520.1 | - | 2289 | ||||
BC031384.1 | - | 2 | 192 | 5356491 | ||
BC086880.1 | - | 2 | 155 | 6762989 | ||
BC089555.1 | - | 2 | 195 | 30312783 | ||
BC094407.1 | - | 2 | 149 | 573475 | ||
BY269662.1 | - | 2 | 220 | |||
Y17223.1 | - | 2 | 149 |
