siRNA Details
Gene Symbol: LOC681067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257516.2 | XP_006257578.1 | 1 | 706 |
Gene Symbol: LOC103690904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773582.1 | XP_008771804.1 | 1 | 497 |
Gene Symbol: LOC691030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773536.1 | XP_008771758.1 | 1 | 635 |
Gene Symbol: RGD1566225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257564.2 | XP_006257626.1 | 1 | 497 |
