siRNA Details
Gene Symbol: AP4S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007077.4 | NP_009008.2 | 5, 6 | 748 | ||
XM_005267293.4 | XP_005267350.1 | 6, 7 | 1191 | |||
XM_011536371.2 | XP_011534673.1 | 6, 7 | 761 | |||
XM_011536372.2 | XP_011534674.1 | 5, 6 | 1155 | |||
GenBank mRNA | BC001259.1 | - | 5, 6 | 531 | 3454117 | |
BT006701.1 | - | 4, 5 | 359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
