siRNA Details
Gene Symbol: Ppp3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017042.2 | NP_058738.1 | 1 | 296 | ||
GenBank mRNA | D90036.1 | - | 1 | 296 | ||
M31809.1 | - | 1 | 296 |

More Information
Other Species Targeted

Gene Ontology Categories:
