siRNA Details
Gene Symbol: Cartpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017110.1 | NP_058806.1 | 3 | 584 | ||
XM_006231843.2 | XP_006231905.1 | 3 | 969 | |||
GenBank mRNA | U10071.1 | - | 3 | 584 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
