siRNA Details
Gene Symbol: Crygc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081660.1 | NP_001075129.1 | 3 | 538 | ||
NM_033483.1 | NP_264077.1 | 2 | 521 |

More Information
Other Species Targeted

Gene Ontology Categories:
