siRNA Details
Gene Symbol: LOC100910829 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236799.2 | XP_006236861.1 | 3 | 868 | ||
XM_006236800.2 | XP_006236862.1 | 867 | ||||
XM_006236801.2 | XP_006236863.1 | 853 |
Gene Symbol: Cml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173449.1 | NP_001166920.1 | 1 | 596 | ||
XM_006236785.2 | XP_006236847.1 | 3 | 957 | |||
XM_006236786.2 | XP_006236848.1 | 3 | 983 | |||
XM_006236787.2 | XP_006236849.1 | 3 | 967 |
