siRNA Details
Gene Symbol: Ly6i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139257.1 | NP_640350.1 | 3 | 221 | ||
XM_006241765.2 | XP_006241827.1 | 208 | ||||
XM_006241766.2 | XP_006241828.1 | 4 | 348 | |||
XM_008765545.1 | XP_008763767.1 | 4 | 335 | |||
GenBank mRNA | M30689.1 | - | 3 | 221 |
