siRNA Details
Gene Symbol: Ptpn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145683.1 | NP_663716.1 | 5 | 549 | ||
XM_006249820.2 | XP_006249882.1 | 5 | 1024 | |||
XM_006249821.2 | XP_006249883.1 | 5 | 679 | |||
GenBank mRNA | U28356.1 | - | 5 | 549 |
