siRNA Details
Gene Symbol: Irf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047086.1 | NP_001040551.1 | 6, 7 | 644 | ||
XM_006253139.2 | XP_006253201.1 | 6, 7 | 905 | |||
XM_008771256.1 | XP_008769478.1 | 6, 7 | 740 | |||
GenBank mRNA | BC166593.1 | - | 6, 7 | 685 | 7934031 | |
DQ674266.1 | - | 6, 7 | 644 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008391, AK155025, BC006577, BC110666, AK036652, and others.
Gene Ontology Categories:
