siRNA Details
Gene Symbol: Whsc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106090.1 | NP_001099560.1 | 2 | 706 | ||
XM_006253307.2 | XP_006253369.1 | 2 | 823 | |||
XM_006253309.2 | XP_006253371.1 | 2 | 823 | |||
XM_006253311.2 | XP_006253373.1 | 2 | 823 | |||
XM_008771349.1 | XP_008769571.1 | 2 | 2694 | |||
XM_008771350.1 | XP_008769572.1 | 2 | 822 |
