siRNA Details
Gene Symbol: LOC102553316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235107.2 | XP_006235169.2 | 1 | 324 |
Gene Symbol: Ssty1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_229292.8 | XP_229292.6 | 1 | 324 |
