siRNA Details
Gene Symbol: Spry4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106150.1 | NP_001099620.1 | 2 | 454 | ||
XM_008772067.1 | XP_008770289.1 | 2 | 1311 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011898, AK048122, BC050944, AK037180, AB019280, and others.
Gene Ontology Categories:
