siRNA Details
Gene Symbol: Ethe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106234.1 | NP_001099704.1 | 5 | 629 | ||
GenBank mRNA | BC158848.1 | - | 5 | 589 | 7444454 | |
FQ209618.1 | - | 5 | 600 | |||
FQ221207.1 | - | 1 | 41 |
