siRNA Details
Gene Symbol: Usp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233262.1 | XP_006233324.1 | 7 | 1342 | ||
GenBank mRNA | FQ222386.1 | - | 7 | 1347 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Usp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233262.1 | XP_006233324.1 | 7 | 1342 | ||
GenBank mRNA | FQ222386.1 | - | 7 | 1347 |