siRNA Details
Gene Symbol: Sestd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134514.2 | NP_001127986.2 | 17 | 1971 | ||
XM_006234402.2 | XP_006234464.1 | 18 | 2251 | |||
XM_006234404.2 | XP_006234466.1 | 17 | 2161 | |||
XM_008761892.1 | XP_008760114.1 | 18 | 2177 | |||
XM_008761893.1 | XP_008760115.1 | 17 | 2087 | |||
GenBank mRNA | BC169110.1 | - | 17 | 1971 | 7445081 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
