siRNA Details
Gene Symbol: Csgalnact2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106616.1 | NP_001100086.1 | 2 | 917 | ||
XM_006237182.2 | XP_006237244.1 | 2 | 931 | |||
GenBank mRNA | FQ221326.1 | - | 2 | 829 |
