siRNA Details
Gene Symbol: Chmp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025410.1 | NP_001020581.1 | 2, 3 | 219 | ||
GenBank mRNA | BC086333.1 | - | 1, 2 | 81 | ||
BC097963.1 | - | 2, 3 | 219 | 7387550 | ||
FQ214132.1 | - | 2, 3 | 247 | |||
FQ228147.1 | - | 2, 3 | 253 | |||
FQ234681.1 | - | 2, 3 | 253 |

More Information
Other Species Targeted

Gene Ontology Categories:
