siRNA Details
Gene Symbol: Rad23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025275.1 | NP_001020446.1 | 9 | 1394 | ||
GenBank mRNA | BC070960.1 | - | 5 | 613 | ||
BC090351.1 | - | 6 | 857 | |||
BC098674.1 | - | 9 | 1394 | 7190031 | ||
BC111406.1 | - | 4 | 533 |
