siRNA Details
Gene Symbol: Ipp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106679.2 | NP_001100149.2 | 2 | 424 | ||
XM_006238640.2 | XP_006238702.1 | 3 | 596 | |||
XM_006238642.2 | XP_006238704.1 | 3 | 607 | |||
XM_006238643.2 | XP_006238705.1 | 3 | 608 | |||
XM_006238644.2 | XP_006238706.1 | 3 | 608 | |||
XM_006238645.2 | XP_006238707.1 | 3 | 608 | |||
XM_008763992.1 | XP_008762214.1 | 3 | 607 |
