siRNA Details
Gene Symbol: Dmap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015006.1 | NP_001015006.1 | 4, 5 | 500 | ||
GenBank mRNA | BC092651.1 | - | 4, 5 | 500 | 7319534 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dmap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015006.1 | NP_001015006.1 | 4, 5 | 500 | ||
GenBank mRNA | BC092651.1 | - | 4, 5 | 500 | 7319534 |