siRNA Details
Gene Symbol: Gpalpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024875.1 | NP_001020046.1 | 6 | 702 | ||
GenBank mRNA | BC097485.1 | - | 6 | 702 | 7459542 | |
FQ214079.1 | - | 390 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpalpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024875.1 | NP_001020046.1 | 6 | 702 | ||
GenBank mRNA | BC097485.1 | - | 6 | 702 | 7459542 | |
FQ214079.1 | - | 390 |