siRNA Details
Gene Symbol: Fkbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106736.2 | NP_001100206.2 | 6 | 560 | ||
GenBank mRNA | BC168873.1 | - | 6 | 525 | ||
FQ214685.1 | - | 2 | 78 | |||
FQ214789.1 | - | 2 | 78 | |||
FQ223884.1 | - | 2 | 78 |
