siRNA Details
Gene Symbol: Fkbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106736.2 | NP_001100206.2 | 5 | 511 | ||
GenBank mRNA | BC168873.1 | - | 5 | 476 | ||
FQ214685.1 | - | 1 | 29 | |||
FQ214789.1 | - | 1 | 29 | |||
FQ223884.1 | - | 1 | 29 |
