siRNA Details
Gene Symbol: Rgl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106805.1 | NP_001100275.1 | 14, 15 | 2027 | ||
XM_006242630.2 | XP_006242692.2 | 17, 18 | 2352 |

More Information
Other Species Targeted

Gene Ontology Categories:
