siRNA Details
Gene Symbol: Tbx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106937.1 | NP_001100407.1 | 3 | 335 | ||
XM_006257159.2 | XP_006257221.1 | 4 | 604 | |||
XM_006257160.2 | XP_006257222.1 | 4 | 454 | |||
XM_006257161.2 | XP_006257223.1 | 4 | 601 | |||
XM_006257162.2 | XP_006257224.1 | 3 | 644 | |||
XM_008773339.1 | XP_008771561.1 | 3 | 319 |

More Information
Other Species Targeted

Gene Ontology Categories:
