siRNA Details
Gene Symbol: Stxbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107038.1 | NP_001100508.1 | 10, 11 | 1038 | ||
XM_006247128.2 | XP_006247190.1 | 10, 11 | 1154 | |||
XM_006247129.2 | XP_006247191.1 | 10, 11 | 1137 | |||
XM_006247131.2 | XP_006247193.1 | 10, 11 | 1154 | |||
XM_008768042.1 | XP_008766264.1 | 11, 12 | 1246 | |||
XM_008768043.1 | XP_008766265.1 | 10, 11 | 1154 |
