siRNA Details
Gene Symbol: RGD1563746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247481.2 | XP_006247543.1 | 5 | 2429 | ||
XM_220997.8 | XP_220997.5 | 5 | 2489 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001127576, CJ046726.
