siRNA Details
Gene Symbol: Thap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107209.2 | NP_001100679.1 | 4 | 390 | ||
XM_008770030.1 | XP_008768252.1 | 4 | 578 | |||
XM_008770031.1 | XP_008768253.1 | 3 | 326 | |||
XM_008770032.1 | XP_008768254.1 | 3 | 329 | |||
XM_008770033.1 | XP_008768255.1 | 3 | 331 | |||
XM_008770034.1 | XP_008768256.1 | 2 | 228 | |||
XM_008770035.1 | XP_008768257.1 | 3 | 270 | |||
GenBank mRNA | CK602350.1 | - | 3 | 289 | ||
FM098768.1 | - | 4 | 390 |
