siRNA Details
Gene Symbol: Epc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728490.4 | XP_002728536.1 | 7 | 1194 | ||
XM_003751704.2 | XP_003751752.1 | 7 | 1301 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
