siRNA Details
Gene Symbol: Fam120b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107466.3 | NP_001100936.2 | 8 | 2584 | ||
XM_006231141.2 | XP_006231203.1 | 8 | 2713 | |||
XM_006231142.2 | XP_006231204.1 | 7 | 2619 | |||
XM_008758761.1 | XP_008756983.1 | 8 | 2425 | |||
GenBank mRNA | FQ210918.1 | - | 8 | 2584 |
