siRNA Details
Gene Symbol: LOC102552284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758851.1 | XP_008757073.1 | 4 | 3022 | ||
XM_008758852.1 | XP_008757074.1 | 4 | 1681 | |||
XM_008758853.1 | XP_008757075.1 | 2 | 1533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
